Assay Details
| Gene Symbol: LOC100616526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038195.2 | 2 - 3 | 267 | 76 | ||
| GenBank mRNA | DV832892.1 | - | 2 - 3 | 268 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-9-2 | MIR9-2 |
bta-mir-9-2, mir-9-2 |
-4.1kb | Sense | Chr. 7 - 88196006 - 88196092 on NCBI Build ARS-UCD1.2 |
| Downstream | myocyte enhancer factor 2C | MEF2C | 44.4kb | Sense | Chr. 7 - 88246676 - 88425829 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

LOC100616526
Genomic Map