Assay Details
| Gene Symbol: MIR103-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031265.1 | 1 | 34 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | spermatogenic leucine zipper 1 | SPZ1 | -28.8kb | Anti-sense | Chr. 20 - 276052 - 283737 on NCBI Build ARS-UCD1.2 | |
| Downstream | slit guidance ligand 3 | SLIT3 | 130.4kb | Sense | Chr. 20 - 442969 - 1163718 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR103-1
Genomic Map