Assay Details
| Gene Symbol: MIR767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031381.1 | 1 | 60 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-105a | MIR105A | bta-mir-105a | -1.4kb | Sense | Chr. X - 34766804 - 34766884 on NCBI Build ARS-UCD1.2 |
| Downstream | microRNA mir-105b | MIR105B | bta-mir-105b | 833.0 | Sense | Chr. X - 34769258 - 34769337 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR767
Genomic Map