Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302662.1 | NP_001289591.1 | 2 - 3 | 397 | 70 | |
| GenBank mRNA | CK778655.1 | - | 2 - 3 | 649 | 70 | |
More Information
Set Membership: |
|

SLAMF9
Genomic Map