Assay Details
| Gene Symbol: TMEM200B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302948.2 | NP_001289877.1 | 2 - 3 | 194 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302948.2 transcripts.
Set Membership: |
|

TMEM200B
Genomic Map