Assay Details
| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303435.1 | NP_001290364.1 | 3 - 4 | 281 | 58 | |
| GenBank mRNA | HQ697269.1 | - | 3 - 4 | 281 | 58 | |
More Information
Set Membership: |
|

TNNI1
Genomic Map