Assay Details
| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC148922.1 | - | 4 - 5 | 1721 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF543| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC148922.1 | - | 4 - 5 | 1721 | 116 | |
Set Membership: |
|