Assay Details
| Gene Symbol: TADA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206410.1 | NP_001193339.1 | 1 - 2 | 309 | 112 | |
| GenBank mRNA | DT900345.1 | - | 1 - 2 | 309 | 112 | |
More Information
Set Membership: |
|

TADA2B
Genomic Map