Assay Details
| Gene Symbol: GPR39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143744.1 | NP_001137216.1 | 2 | 2317 | 72 | |
| GenBank mRNA | EU031892.1 | - | 2 | 2317 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR39| Gene Symbol: GPR39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143744.1 | NP_001137216.1 | 2 | 2317 | 72 | |
| GenBank mRNA | EU031892.1 | - | 2 | 2317 | 72 | |
Set Membership: |
|