Assay Details
| Gene Symbol: tap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078027.6 | NP_510428.1 | 7 - 8 | 940 | 77 | |
| GenBank mRNA | AF145375.1 | - | 6 - 7 | 734 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tap-1| Gene Symbol: tap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078027.6 | NP_510428.1 | 7 - 8 | 940 | 77 | |
| GenBank mRNA | AF145375.1 | - | 6 - 7 | 734 | 77 | |
Set Membership: |
|