Assay Details
| Gene Symbol: nfki-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078139.5 | NP_510540.1 | 3 - 4 | 511 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nfki-1| Gene Symbol: nfki-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078139.5 | NP_510540.1 | 3 - 4 | 511 | 60 | |
Set Membership: |
|