Assay Details
| Gene Symbol: gcy-36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078156.8 | NP_510557.3 | 7 - 8 | 965 | 117 | |
| GenBank mRNA | AY652945.1 | - | 7 - 8 | 815 | 117 | |
More Information
Set Membership: |
|

gcy-36
Genomic Map