Assay Details
| Gene Symbol: pbo-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307904.3 | NP_001294833.1 | 3 - 4 | 370 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pbo-6| Gene Symbol: pbo-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307904.3 | NP_001294833.1 | 3 - 4 | 370 | 106 | |
Set Membership: |
|