Assay Details
| Gene Symbol: fhod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313484.2 | NP_001300413.1 | 15 - 16 | 3511 | 79 | |
| NM_001313485.3 | NP_001300414.1 | 17 - 18 | 4035 | 79 | ||
| NM_001313486.4 | NP_001300415.1 | 16 - 17 | 3564 | 79 | ||
| NM_001313487.3 | NP_001300416.1 | 16 - 17 | 5040 | 79 | ||
| NM_001313488.4 | NP_001300417.1 | 15 - 16 | 4993 | 79 | ||
| NM_001313489.3 | NP_001300418.1 | 15 - 16 | 4566 | 79 | ||
| NM_001313490.3 | NP_001300419.1 | 14 - 15 | 4515 | 79 | ||
| NM_059112.4 | NP_491513.2 | 16 - 17 | 3985 | 79 | ||
| GenBank mRNA | AB084086.1 | - | 16 - 17 | 4007 | 79 | |
More Information
Set Membership: |
|

fhod-1
Genomic Map