Assay Details
| Gene Symbol: fhod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313484.2 | NP_001300413.1 | 8 - 9 | 1185 | 119 | |
| NM_001313485.3 | NP_001300414.1 | 10 - 11 | 1709 | 119 | ||
| NM_001313486.4 | NP_001300415.1 | 9 - 10 | 1238 | 119 | ||
| NM_001313487.3 | NP_001300416.1 | 9 - 10 | 2714 | 119 | ||
| NM_001313488.4 | NP_001300417.1 | 8 - 9 | 2667 | 119 | ||
| NM_001313489.3 | NP_001300418.1 | 8 - 9 | 2240 | 119 | ||
| NM_001313490.3 | NP_001300419.1 | 7 - 8 | 2189 | 119 | ||
| NM_059112.4 | NP_491513.2 | 9 - 10 | 1659 | 119 | ||
| GenBank mRNA | AB084086.1 | - | 9 - 10 | 1681 | 119 | |
More Information
Set Membership: |
|

fhod-1
Genomic Map