Assay Details
| Gene Symbol: kin-32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170854.5 | NP_740841.1 | 3 - 4 | 317 | 86 | |
| NM_170855.6 | NP_740842.1 | 3 - 4 | 399 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kin-32| Gene Symbol: kin-32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170854.5 | NP_740841.1 | 3 - 4 | 317 | 86 | |
| NM_170855.6 | NP_740842.1 | 3 - 4 | 399 | 86 | ||
Set Membership: |
|