Assay Details
| Gene Symbol: kin-32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170854.5 | NP_740841.1 | 5 - 6 | 593 | 105 | |
| NM_170855.6 | NP_740842.1 | 5 - 6 | 675 | 105 | ||
More Information
Set Membership: |
|

kin-32
Genomic Map