Assay Details
| Gene Symbol: kin-32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170854.5 | NP_740841.1 | 7 - 8 | 1896 | 86 | |
| NM_170855.6 | NP_740842.1 | 7 - 8 | 1978 | 86 | ||
More Information
Set Membership: |
|

kin-32
Genomic Map