Assay Details
| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059957.5 | NP_492358.3 | 10 - 11 | 1482 | 117 | |
| NM_170904.3 | NP_740896.2 | 10 - 11 | 1482 | 117 | ||
| NM_170905.3 | NP_740897.1 | 4 - 5 | 492 | 117 | ||
More Information
Set Membership: |
|

cle-1
Genomic Map