Assay Details
| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170904.3 | NP_740896.2 | 14 - 15 | 1923 | 149 | |
| NM_170905.3 | NP_740897.1 | 8 - 9 | 933 | 149 | ||
| NM_170906.6 | NP_740898.1 | 2 - 3 | 459 | 149 | ||
More Information
Set Membership: |
|

cle-1
Genomic Map