Assay Details
| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059957.5 | NP_492358.3 | 20 - 21 | 3270 | 106 | |
| NM_170904.3 | NP_740896.2 | 18 - 19 | 3159 | 106 | ||
| NM_170905.3 | NP_740897.1 | 12 - 13 | 2169 | 106 | ||
| NM_170906.6 | NP_740898.1 | 6 - 7 | 1695 | 106 | ||
More Information
Set Membership: |
|

cle-1
Genomic Map