Assay Details
| Gene Symbol: cle-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059957.5 | NP_492358.3 | 7 - 8 | 1190 | 134 | |
| NM_170904.3 | NP_740896.2 | 7 - 8 | 1190 | 134 | ||
More Information
Set Membership: |
|

cle-1
Genomic Map