Assay Details
| Gene Symbol: pmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026691.5 | NP_001021862.1 | 10 - 11 | 2815 | 96 | |
| NM_001373648.3 | NP_001359606.1 | 10 - 11 | 2841 | 96 | ||
| GenBank mRNA | AJ303081.1 | - | 10 - 11 | 2869 | 96 | |
| AJ303082.1 | - | 10 - 11 | 2863 | 96 | ||
More Information
Set Membership: |
|

pmr-1
Genomic Map