Assay Details
| Gene Symbol: pmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026691.5 | NP_001021862.1 | 2 - 3 | 141 | 165 | |
| NM_001373648.3 | NP_001359606.1 | 2 - 3 | 167 | 165 | ||
| GenBank mRNA | AJ303081.1 | - | 2 - 3 | 195 | 165 | |
| AJ303082.1 | - | 2 - 3 | 189 | 165 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ303081.1, AJ303082.1, NM_001373648.3 transcripts.
Set Membership: |
|

pmr-1
Genomic Map