Assay Details
| Gene Symbol: pmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026691.5 | NP_001021862.1 | 8 - 9 | 2313 | 90 | |
| NM_001373648.3 | NP_001359606.1 | 8 - 9 | 2339 | 90 | ||
| GenBank mRNA | AJ303081.1 | - | 8 - 9 | 2367 | 90 | |
| AJ303082.1 | - | 8 - 9 | 2361 | 90 | ||
More Information
Set Membership: |
|

pmr-1
Genomic Map