Assay Details
| Gene Symbol: swp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025951.3 | NP_001021122.1 | 3 - 4 | 319 | 112 | |
| NM_001379724.2 | NP_001367326.1 | 4 - 5 | 362 | 112 | ||
| NM_001382877.1 | 4 - 5 | 364 | 112 | |||
| NM_171132.4 | NP_741149.1 | 2 - 3 | 307 | 112 | ||
| GenBank mRNA | U06932.1 | - | 2 - 3 | 307 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379724.2 transcripts.
Set Membership: |
|

swp-1
Genomic Map