Assay Details
| Gene Symbol: clp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379816.2 | NP_001367451.1 | 1 - 2 | 99 | 105 | |
| NM_001379817.1 | NP_001367450.1 | 1 - 2 | 188 | 105 | ||
| NM_066339.6 | NP_498740.2 | 1 - 2 | 194 | 105 | ||
| NM_066340.6 | NP_498741.3 | 2 - 3 | 269 | 105 | ||
More Information
Set Membership: |
|

clp-1
Genomic Map