Assay Details
| Gene Symbol: aak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382945.1 | 6 - 7 | 1463 | 136 | ||
| NM_171216.5 | NP_741254.3 | 6 - 7 | 1464 | 136 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aak-1| Gene Symbol: aak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382945.1 | 6 - 7 | 1463 | 136 | ||
| NM_171216.5 | NP_741254.3 | 6 - 7 | 1464 | 136 | ||
Set Membership: |
|