Assay Details
| Gene Symbol: prdx-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171241.4 | NP_741287.1 | 2 - 3 | 487 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prdx-6| Gene Symbol: prdx-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171241.4 | NP_741287.1 | 2 - 3 | 487 | 69 | |
Set Membership: |
|