Assay Details
| Gene Symbol: spg-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059487.6 | NP_491888.2 | 7 - 8 | 1145 | 89 | |
| Gene Symbol: ucr-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380573.1 | NP_001367866.1 | 8 - 9 | 1648 | 89 | |
More Information
Set Membership: |
|

spg-20
Genomic Map