Assay Details
| Gene Symbol: bag-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059492.7 | NP_491893.1 | 2 - 3 | 243 | 73 | |
| GenBank mRNA | AF095788.1 | - | 1 - 2 | 158 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bag-1| Gene Symbol: bag-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059492.7 | NP_491893.1 | 2 - 3 | 243 | 73 | |
| GenBank mRNA | AF095788.1 | - | 1 - 2 | 158 | 73 | |
Set Membership: |
|