Assay Details
| Gene Symbol: bag-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059492.7 | NP_491893.1 | 3 - 4 | 590 | 105 | |
| GenBank mRNA | AF095788.1 | - | 2 - 3 | 505 | 105 | |
More Information
Set Membership: |
|

bag-1
Genomic Map