Assay Details
| Gene Symbol: mif-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059668.4 | NP_492069.1 | 2 - 3 | 114 | 74 | |
| Gene Symbol: F13G3.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380688.2 | NP_001366779.1 | 3 - 4 | 557 | 74 | |
More Information
Set Membership: |
|

mif-3
Genomic Map