Assay Details
| Gene Symbol: spe-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059694.6 | NP_492095.1 | 6 - 7 | 1201 | 138 | |
| GenBank mRNA | Z14066.1 | - | 1162 | 138 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spe-4| Gene Symbol: spe-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059694.6 | NP_492095.1 | 6 - 7 | 1201 | 138 | |
| GenBank mRNA | Z14066.1 | - | 1162 | 138 | ||
Set Membership: |
|