Assay Details
| Gene Symbol: lars-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026704.5 | NP_001021875.1 | 8 - 9 | 2427 | 149 | |
| NM_001136331.4 | NP_001129803.1 | 5 - 6 | 1212 | 149 | ||
More Information
Set Membership: |
|

lars-2
Genomic Map