Assay Details
| Gene Symbol: taf-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059768.7 | NP_492169.2 | 6 - 7 | 1977 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

taf-5| Gene Symbol: taf-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059768.7 | NP_492169.2 | 6 - 7 | 1977 | 72 | |
Set Membership: |
|