Assay Details
| Gene Symbol: tnsl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374927.3 | NP_001361839.1 | 16 - 17 | 2607 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tnsl-1| Gene Symbol: tnsl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374927.3 | NP_001361839.1 | 16 - 17 | 2607 | 99 | |
Set Membership: |
|