Assay Details
Gene Symbol: lmd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129003.3 | NP_001122475.1 | 4 - 5 | 541 | 78 | |
NM_001129004.3 | NP_001122476.1 | 4 - 5 | 549 | 78 |
Gene Symbol: F43G9.13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059928.5 | NP_492329.1 | 9 - 10 | 2134 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
