Assay Details
| Gene Symbol: spd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060013.7 | NP_492414.1 | 7 - 8 | 2236 | 122 | |
| GenBank mRNA | AY340594.1 | - | 7 - 8 | 2187 | 122 | |
More Information
Set Membership: |
|

spd-2
Genomic Map