Assay Details
| Gene Symbol: prmt-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060035.4 | NP_492436.2 | 8 - 9 | 1727 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prmt-7| Gene Symbol: prmt-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060035.4 | NP_492436.2 | 8 - 9 | 1727 | 99 | |
Set Membership: |
|