Assay Details
| Gene Symbol: tag-72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060273.9 | NP_492674.2 | 5 - 6 | 734 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tag-72| Gene Symbol: tag-72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060273.9 | NP_492674.2 | 5 - 6 | 734 | 73 | |
Set Membership: |
|