Assay Details
| Gene Symbol: coa-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381194.1 | NP_001367155.1 | 9 - 10 | 1363 | 133 | |
| NM_001381195.2 | NP_001366833.1 | 6 - 7 | 1103 | 133 | ||
More Information
Set Membership: |
|

coa-6
Genomic Map