Assay Details
| Gene Symbol: herc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306503.3 | NP_001293432.1 | 9 - 10 | 1350 | 102 | |
| NM_001306504.3 | NP_001293433.1 | 8 - 9 | 1215 | 102 | ||
| NM_001306505.3 | NP_001293434.1 | 6 - 7 | 798 | 102 | ||
| NM_058433.5 | NP_490834.1 | 14 - 15 | 2353 | 102 | ||
More Information
Set Membership: |
|

herc-1
Genomic Map