Assay Details
| Gene Symbol: spe-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306501.3 | NP_001293430.1 | 8 - 9 | 1266 | 65 | |
| NM_001306502.3 | NP_001293431.1 | 4 - 5 | 732 | 65 | ||
| NM_058455.5 | NP_490856.1 | 15 - 16 | 3382 | 65 | ||
More Information
Set Membership: |
|

spe-15
Genomic Map