Assay Details
| Gene Symbol: spe-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058455.5 | NP_490856.1 | 2 - 3 | 369 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spe-15| Gene Symbol: spe-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058455.5 | NP_490856.1 | 2 - 3 | 369 | 104 | |
Set Membership: |
|