Assay Details
| Gene Symbol: spe-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306501.3 | NP_001293430.1 | 1 - 2 | 279 | 62 | |
| NM_058455.5 | NP_490856.1 | 8 - 9 | 2395 | 62 | ||
More Information
Set Membership: |
|

spe-15
Genomic Map