Assay Details
| Gene Symbol: spe-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306501.3 | NP_001293430.1 | 2 - 3 | 396 | 66 | |
| NM_058455.5 | NP_490856.1 | 9 - 10 | 2512 | 66 | ||
More Information
Set Membership: |
|

spe-15
Genomic Map