Assay Details
| Gene Symbol: spe-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306501.3 | NP_001293430.1 | 3 - 4 | 483 | 70 | |
| NM_058455.5 | NP_490856.1 | 10 - 11 | 2599 | 70 | ||
More Information
Set Membership: |
|

spe-15
Genomic Map