Assay Details
| Gene Symbol: afd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026489.7 | NP_001021660.2 | 13 - 14 | 2905 | 60 | |
| NM_001026490.6 | NP_001021661.2 | 13 - 14 | 2903 | 60 | ||
| NM_001306400.4 | NP_001293329.1 | 13 - 14 | 2907 | 60 | ||
| NM_001306401.3 | NP_001293330.1 | 8 - 9 | 2042 | 60 | ||
| NM_001306402.3 | NP_001293331.1 | 8 - 9 | 2042 | 60 | ||
| NM_001306403.3 | NP_001293332.1 | 7 - 8 | 1856 | 60 | ||
| NM_001306404.3 | NP_001293333.1 | 7 - 8 | 1856 | 60 | ||
| NM_001306405.3 | NP_001293334.1 | 4 - 5 | 1577 | 60 | ||
| NM_001306406.3 | NP_001293335.1 | 4 - 5 | 1577 | 60 | ||
| NM_001306407.3 | NP_001293336.1 | 2 - 3 | 1331 | 60 | ||
| NM_001306408.3 | NP_001293337.1 | 2 - 3 | 1331 | 60 | ||
| NM_001306529.4 | NP_001293458.1 | 3 - 4 | 1660 | 60 | ||
| NM_001356800.4 | NP_001343732.1 | 10 | 2458 | 60 | ||
| NM_058537.5 | NP_490938.3 | 13 - 14 | 2899 | 60 | ||
More Information
Set Membership: |
|

afd-1
Genomic Map