Assay Details
| Gene Symbol: afd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026489.7 | NP_001021660.2 | 14 - 15 | 2996 | 85 | |
| NM_001026490.6 | NP_001021661.2 | 14 - 15 | 2994 | 85 | ||
| NM_001306400.4 | NP_001293329.1 | 14 - 15 | 2998 | 85 | ||
| NM_001306401.3 | NP_001293330.1 | 9 - 10 | 2133 | 85 | ||
| NM_001306402.3 | NP_001293331.1 | 9 - 10 | 2133 | 85 | ||
| NM_001306403.3 | NP_001293332.1 | 8 - 9 | 1947 | 85 | ||
| NM_001306404.3 | NP_001293333.1 | 8 - 9 | 1947 | 85 | ||
| NM_001306405.3 | NP_001293334.1 | 5 - 6 | 1668 | 85 | ||
| NM_001306406.3 | NP_001293335.1 | 5 - 6 | 1668 | 85 | ||
| NM_001306407.3 | NP_001293336.1 | 3 - 4 | 1422 | 85 | ||
| NM_001306408.3 | NP_001293337.1 | 3 - 4 | 1422 | 85 | ||
| NM_001306529.4 | NP_001293458.1 | 4 - 5 | 1751 | 85 | ||
| NM_001356800.4 | NP_001343732.1 | 11 | 2549 | 85 | ||
| NM_058537.5 | NP_490938.3 | 14 - 15 | 2990 | 85 | ||
More Information
Set Membership: |
|

afd-1
Genomic Map